Tisk…

Anti-Thioredoxin 2 Mouse Monoclonal Antibody [clone: 4C5]

Dodavatel: BioVision
BIOV6167-100EA 10350 CZK
BIOV6167-100
Anti-Thioredoxin 2 Mouse Monoclonal Antibody [clone: 4C5]
The mammalian thioredoxin reductases (TrxRs) are a family of seleno-cysteine containing pyridine nucleotide-disulfide oxido-reductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate seleno-cysteine (SeCys) residue in the conserved sequence(-Gly-Cys-SeCys-Gly;TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55 – 58 kDa in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR;TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena. Trx 2 Has an anti-apoptotic function and plays an important role in the regulation of mitochondrial membrane potential. Could be involved in the resistance to anti-tumor agents. Possesses a dithiol-reducing activity.

Recommended Dilutions: Western Blot: 1 µg/ml, IP: 2 µg.

Type: Primary
Antigen: Thioredoxin 2
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR