Tisk…

Anti-JWH-250, Synthetic Cannabinoid Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6206-200EA 7470 CZK
BIOV6206-200
Anti-JWH-250, Synthetic Cannabinoid Rabbit Polyclonal Antibody
Protilátky
Cannabinoids are a class of diverse chemical compounds that activate cannabinoid receptors on cells that repress neurotransmitter release in the brain. They are active chemicals in Cannabis that cause drug-like effects throughout the body, including the central nervous system and the immune system. Anti-JWH-250 is a rabbit polyclonal IgG antibody. It has been used in a competitive ELISA format to test the presence of JWH-250 and its metabolites in samples such as urine, whole blood, serum, and plasma (see Arntson et al, 2013;Note: If this antibody is used in an immunoassay to detect synthetic cannabinoids, suspect test samples must be confirmed using an alternative analytical method, for example LC-MS-MS.

Recommended Dilutions: ELISA: for 96-well plate coating use 1-3 ?g/mL;

Type: Primary
Antigen: JWH-250, Synthetic Cannabinoid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR