Tisk…

Anti-BRD3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6282-100EA 7760 CZK
BIOV6282-100
Anti-BRD3 Rabbit Polyclonal Antibody
Protilátky
The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. The BET family of proteins, defined by tandem Bromodomains and an Extra Terminal domain, include BRD2, BRD3, BRD4, and BRDT. The BET proteins play a key role in many cellular processes, including inflammatory gene expression, mitosis, and viral/host interactions. The isolated individual or tandem bromodomains of BRD3 have been shown to bind acetylated histone tails, serving to couple histone acetylation marks to the transcriptional regulation of target promoters. Small molecule inhibitors of these interactions hold promise as useful therapeutics for human disease.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: BRD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR