Tisk…

Anti-Swi6 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6285-100EA 5500 CZK
BIOV6285-100
Anti-Swi6 Rabbit Polyclonal Antibody
Protilátky
Swi6 protein of fission yeast is a functional and structural homolog of HP1 (Heterochromatin Protein 1) of animals and is involved in the formation of heterochromatin structure by binding to centromere, telomere and silent mating-type locus. It is also involved in silencing the genes and sister chromatid cohesion by binding to histone H3 methylated at Lys9 and the cohesin subunit Psc3. It is also involved in the repression of the silent mating-type loci MAT2 and MAT3. It may compact MAT2/3 into a heterochromatin-like conformation which represses the transcription of these silent cassettes.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: Swi6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR