Tisk…

Anti-IgG Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6402-05EA 6350 CZK
BIOV6402-05
Anti-IgG Rabbit Polyclonal Antibody
Protilátky
Rabbit anti-mouse IgG (H+L), horseradish Peroxidase (HPR) conjμgated. This antibody has been optimized as secondary antibody in chemiluminescent Western blotting applications.

Recommended Dilutions: Western blotting: 1:2,000-1:100,000. Enzyme immunoassays: 1:2,000-1:100,000. Immunohistochemistry: 1:200-1:2,000. The optimal conditions should be determined individually.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR