Tisk…

Anti-CETP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6606-200EA 7470 CZK
BIOV6606-200
Anti-CETP Rabbit Polyclonal Antibody
Protilátky
It has been proposed that high density lipoproteins (HDLs) function along with Lecithin, cholesterol acyltransferase and CETP to facilitate cholesterol transport from tissues to the liver. CETP is responsible for neutral lipid transfer activity in plasma in numerous species. Since CETP is able to accelerate specifically the exchange of lipid components between pro and anti-atherogenic lipoprotein fractions, it may be a key determinant of the global atherogenicity of the plasma lipoprotein profile and arises as a possible target in atherosclerosis prevention. CETP has an important role in reverse cholesterol transport and shaping and affecting the composition of plasma lipoproteins. In general elevated levels of CETP have been associated with increased risk of coronary heart disease.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: CETP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR