Tisk…

Anti-Irisin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6635-50EA 9170 CZK
BIOV6635-50
Anti-Irisin Rabbit Polyclonal Antibody
Protilátky
Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin;Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.

Recommended Dilutions: Western Blot - 1: 1000 dilution

Type: Primary
Antigen: Irisin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR