Tisk…

Anti-Galectin-9 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3827-100EA 7780 CZK
BIOV3827-100
Anti-Galectin-9 Rabbit Polyclonal Antibody
Galectin-9 is an eosinophile chemoattractant produced by activated T lymphocytes. It is an integral membrane protein that plays a role in immuno/inflammation processes that causes inflammatory reactions in the central nervous system (CNS;Galectin-9 exists as two isoforms, a long form and a short form, which differ by an internal stretch of 32 amino acids.

Recommended Dilutions: Western Blot: 0.5-4.0 µg/ml)

Type: Primary
Antigen: Galectin-9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR