Tisk…

Anti-Matrilin-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3840-100EA 6910 CZK
BIOV3840-100
Anti-Matrilin-2 Rabbit Polyclonal Antibody
Protilátky
Matrilin-2 is part of the Matrilin family of secreted extracelluar matrix proteins that consist of Matrilin -1 to Matrilin-4. Matrilin-2 encodes a member of the von Willebrand factor A domain protein family. It is involved in matrix assembly and development of filamentous networks in the extracellular matrices of various tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Matrilin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR