Tisk…

Anti-LDH Goat Polyclonal Antibody

Dodavatel: BioVision
BIOV3842-500EA 6350 CZK
BIOV3842-500
Anti-LDH Goat Polyclonal Antibody
Protilátky
Lactate dehydrogenase (LDH) contains two isoforms in mammals, LDH-M and LDH-H, which come together to form a homotetramer of 36 kDa subunits. LDH is often as a marker for tissue breakdown or cytotoxicity. LDH shows cytoplasm localization.

Recommended Dilutions: Western Blot: 1:500-2500

Type: Primary
Antigen: LDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR