Tisk…

Anti-ECM Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3843-100EA 6910 CZK
BIOV3843-100
Anti-ECM Rabbit Polyclonal Antibody
Protilátky
Stearoyl-CoA desaturase (SCD) is a regulatory enzyme involved in the synthesis of oleate and palmitoleate. Oleate and palmitoleate are the main components of membrane phospholipid, triglycerides and cholesterol esters. SCD plays an important role in cellular cholesterol homeostasis and also in the secretion processes of triacylglycerol and phospholipid. Inhibition of SCD could be a useful treatment for metabollic disorders such as obesity and hepatic steatosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ECM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR