Tisk…

Anti-DMP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3844-100EA 7900 CZK
BIOV3844-100
Anti-DMP1 Rabbit Polyclonal Antibody
Protilátky
DMP-1 (Dentin matrix protein1) is a member of the integrin ligand N-linked glycoprotein family.It is mostly found in bone, tooth and hypertrophic cartilage. DMP-1 plays an important role in dentin and bone mineralization. DMP-1 has multiple serine, glutamic acid and aspartic acid residues, a GAG attachement site and RGD motif. DMP-1 is proteolytically proceesed into mulitple fragemtns in the extracellular matrix (ECM) of bone and dentin.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR