Tisk…

Anti-Luciferase Goat Polyclonal Antibody

Dodavatel: BioVision
BIOV3848-500EA 8120 CZK
BIOV3848-500
Anti-Luciferase Goat Polyclonal Antibody
The Firefly luciferase is a genetically modified variant derived from the Luciferase of Diaphanes pectinealis endemic to Yunnan province, China. It has been modified to provide enhanced stability compared to the normal phenotype of Photinus pyralis as well as a broader and more physiologically relevant effective pH range. Luciferase has been used as a sensitive reporter for studying gene regulation and function. Firefly luciferase catalyzes the oxidative carboxylation of luciferin, a reaction with the highest efficiency of any known bioluminescence reaction.

Recommended Dilutions: Western Blot: 1:5000-20,000; ELISA: 1:50,000-200,000; and Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Luciferase
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR