Tisk…

Anti-Proteinase Inhibitor 6 Mouse Monoclonal Antibody [clone: 3A]

Dodavatel: BioVision
BIOV3849-100EA 8320 CZK
BIOV3849-100
Anti-Proteinase Inhibitor 6 Mouse Monoclonal Antibody [clone: 3A]
Protilátky
Proteinase inhibitor 6 (PI-6) is a 42 kDa cytoplasmic protein belonging to the serine proteinase inhibitor (serpin) super family and the ovalbumin-serpin super family. Sequence similarity sμggests that PI-6 most resembles the ovalbumin serpins . Highest levels of PI-6 are found in skeletal muscle. PI-6 is also found in placenta, cardiac muscle, lung, liver, kidney and pancreas.

Recommended Dilutions:

Type: Primary
Antigen: Proteinase Inhibitor 6
Clonality: Monoclonal
Clone: 3A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR