Tisk…
Anti-Beta-Actin Rabbit Polyclonal Antibody
Dodavatel: BioVision
BIOV3850-100EA 8120 CZK
BIOV3850-100
Anti-Beta-Actin Rabbit Polyclonal Antibody
Actin is expressed in all eukaryotic cells and is the major component of the cytoskeleton. At least six types of actin are present in mammalian tissues and fall into three classes. Alpha actin expression is limited to various types of muscle and it regulates contactile potentials for the muscle cells, whereas beta and gamma actin, also known as cytoplasmic actin, are predominantly expressed in nonmuscle cells, controlling cell structure and motility.
Recommended Dilutions: Western Blot: 0.5-2 µg/ml
Type: Primary
Antigen: Beta-Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat