Tisk…

Anti-Ficolin-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3852-100EA 6910 CZK
BIOV3852-100
Anti-Ficolin-1 Rabbit Polyclonal Antibody
Protilátky
Ficolin-1 also known as M-Ficolin is a pattern recognition molecule of the lectin complement activation pathway. It is expressed mostly in monocytes, nuetrophils and in type II alveolar epithelial cells. Ficolin-1 is detected on the surface of circulating monocytes but is not present in the plasma. Ficolin-1 has a 28 aa signal sequence, an N-terminal collagen domain and a C-terminal fibrinogen-like (FBG) domain that contain a calcium binding site and a potential N-glycosylation site.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Ficolin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR