Tisk…

Anti-Neuropilin-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3856-100EA 6910 CZK
BIOV3856-100
Anti-Neuropilin-1 Rabbit Polyclonal Antibody
Protilátky
Neuropilin 1 is a type 1 membrane-bound coreceptor to a tyrosine kinase receptor for both Vascular Endothelial Growth Factor and Semaphorin family members. Neuropilin 1 is invovled in axon growth and guidance, cell survival, migration, and invasion.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Neuropilin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR