Tisk…

Anti-IL-10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6638-100EA 8740 CZK
BIOV6638-100
Anti-IL-10 Rabbit Polyclonal Antibody
IL-10 (Interleukin-10), originally known as Cytokine Synthesis Inhibitory Factor (CSIF), is an 20.5 kDa protein containing 161-178 amino acid residues. IL10 is produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: IL-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR