Tisk…

Anti-TDP2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6697-50EA 11930 CZK
BIOV6697-50
Anti-TDP2 Rabbit Polyclonal Antibody
The TTRAP/TDP2 gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.

  • Recommended Dilutions: Western Blot: ~0.5 µg/ml, ELISA: ~1:62,500 dilution, IHC.

Type: Primary
Antigen: TDP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR