Tisk…

Anti-TDP1 Mouse Monoclonal Antibody [clone: AT1F2]

Dodavatel: BioVision
BIOV6698-50EA 10010 CZK
BIOV6698-50
Anti-TDP1 Mouse Monoclonal Antibody [clone: AT1F2]
Tyrosyl-DNA phosphodiesterase (TDP1) is a DNA repair enzyme that catalyzes the hydrolysis of a phophodiester bond between a tyrosine residue and a DNA 3' phosphate. This protein is a member of the phospholipase D (PLD) superfamily and contains two PLD phosphodiesterase domains. Mutations in TDP1 are associated with the disease spino-cerebellar ataxia with axonal neuropathy (SCAN1) by interfering with DNA transcription or by inducting apoptosis in postmitotic neurons.

Recommended Dilutions: Western Blot: ~1:1000 - 2000, ELISA.

Type: Primary
Antigen: TDP1
Clonality: Monoclonal
Clone: AT1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1k
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR