Tisk…

Anti-FTO Mouse Monoclonal Antibody [clone: FT86-4]

Dodavatel: BioVision
BIOV6699-50EA 8260 CZK
BIOV6699-50
Anti-FTO Mouse Monoclonal Antibody [clone: FT86-4]
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

Recommended Dilutions: ELISA, IP, Western Blot

Type: Primary
Antigen: FTO
Clonality: Monoclonal
Clone: FT86-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1k
Reactivity: Human, Rat And Mouse.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR