Tisk…

Anti-ROR2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6700-100EA 7780 CZK
BIOV6700-100
Anti-ROR2 Rabbit Polyclonal Antibody
ROR2 (receptor tyrosine kinase-like orphan receptor 2), also known as neurotrophic tyrosine kinase receptor-related 2 (NTRKR2), is a single pass transmembrane tyrosine-protein kinase receptor. It contains a cytoplasmic tyrosine kinase domain, distally located serine-threonine-rich domains, an extracellular immunoglobulin-like domain, a cysteine-rich domain and a kringle domain. ROR2 is important for skeletal and endocrine development and is required for cartilage and growth plate development. It promotes the differentiation of osteoblasts and plays an important role in the early formation of chondrocytes. ROR2 may play differential roles during the development of the nervous system. ROR2 sequesters and associates with Dlxin-1 affecting the transcriptional function of Msx-2. ROR2 also interacts with canonical Wnt1 and Wnt3, regulating their signaling pathways. Defects in ROR2 can result in the autosomal dominant skeletal disorder, brachydactylic type B1 or the autosomal recessive skeletal disorder, Robinow syndrome.

Recommended Dilutions: Western Blot: 1:1000, IHC: 1:10 to 1:50, FACS: 1:10 to 1:50

Type: Primary
Antigen: ROR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR