Tisk…

Anti-AGT Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6708-100EA 6910 CZK
BIOV6708-100
Anti-AGT Rabbit Polyclonal Antibody
Protilátky
Angiotensinogen, also known as AGT and SERPINA8, is a member of the serpin family. It is anα-2-globulinthat is produced constitutively and released into the circulation mainly by the liver. Angiotensinogen is a essential component of the renin-angiotensin system (RAS) and a potent regulator of blood pressure. It is expressed by the liver and secreted in plasma. Angiotensinogen is cleaved into three chains: Angiotensin-1 (Ang I), Angiotensin-2 (Ang II), and Angiotensin-3 (Ang III;Angiotensin-1 is a substrate of ACE (angiotensin converting enzyme) that removes a dipeptide to yield the physiologically active peptide angiotensin-2. Angiotensin-1 and angiotensin-2 can be further processed to generate angiotensin-3, angiotensin-4. Angiotensin 1-7 is cleaved from angiotensin-2 by ACE2. Angiotensin-2 acts directly on vascular smooth muscle as a potent vasoconstrictor, affects cardiac contractility and heart rate through its action on the sympathetic nervous system. Defects in AGT are associated with susceptibility to essential hypertension and renal tubular dysgenesis (RTD).

Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:10-1:50

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR