Tisk…

Anti-Androgen Receptor Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6710-100EA 7780 CZK
BIOV6710-100
Anti-Androgen Receptor Rabbit Polyclonal Antibody
Androgens exhibit a wide range of effects on the development, maintenance and regulation of male phenotype and reproductive physiology in males. The androgen receptor (AR) is a member of the steroid superfamily of ligand-dependent transcription factors. ARs bind active testosterone (T) and dihydrotestosterone (DHT;The rates of association and dissociation of T are about 3 times more rapid than those of DHT. This difference in binding kinetics may account for the different physiological effects of T and DHT. Androgen binding results in an at least 6-fold increase in androgen receptor stability.

Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:10-1:50

Type: Primary
Antigen: Androgen Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR