Tisk…

Anti-His-Tag Mouse Monoclonal Antibody [clone: 11E12]

Dodavatel: BioVision
BIOV6714-100EA 10970 CZK
BIOV6714-100
Anti-His-Tag Mouse Monoclonal Antibody [clone: 11E12]
A variety of plasmids contain DNA that encodes an N-terminal tag consisting of six histidine (His) residues, followed by an extended multiple cloning sites. The 6xHis tag on the expressed recombinant proteins allows for efficient coupling to Ni++ affinity resins and purification by a single step chromatography. As is the case with other protein tags, this polyhistidine tag can be cleaved at sites recognized by enzymes such as thrombin and enterokinases to isolate the protein of interest.

Recommended Dilutions: ELISA, WB

Type: Primary
Antigen: His-Tag
Clonality: Monoclonal
Clone: 11E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR