Tisk…

Anti-ApoA5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6717-100EA 6910 CZK
BIOV6717-100
Anti-ApoA5 Rabbit Polyclonal Antibody
Protilátky
Apolipoproteins are plasma lipoproteins that function as transporters of lipids and cholesterol in the circulatory system. The protein encoded by this gene is an apolipoprotein that plays an important role in regulating the plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. Mutations in this gene have been associated with hypertriglyceridemia and hyperlipoproteinemia type 5.

Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:10-1:50.

Type: Primary
Antigen: ApoA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR