Tisk…

Anti-ApoD Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6719-100EA 7780 CZK
BIOV6719-100
Anti-ApoD Rabbit Polyclonal Antibody
Apolipoproteins are plasma lipoproteins that function as transporters of lipids and cholesterol in the circulatory system. Apolipoprotein-D (Apo-D) is mainly associated with high density lipoproteins in human plasma. It is expressed in numerous tissues having high levels of expression in spleen, testes and brain. Apo-D is a multi-ligand, multi-functional transporter and transports a ligand from 1 cell to another within an organ, scavenge a ligand within an organ for transport to the blood or could transport a ligand from the circulation to specific cells within a tissue.

Recommended Dilutions: Western Blot: ~1:1000.

Type: Primary
Antigen: ApoD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR