Tisk…

Anti-SCD Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3875-100EA 6910 CZK
BIOV3875-100
Anti-SCD Rabbit Polyclonal Antibody
Protilátky
Stearoyl-CoA desaturase (SCD) is a regulatory enzyme involved in the synthesis of oleate and palmitoleate. Oleate and palmitoleate are the main components of membrane phospholipid, triglycerides and cholesterol esters. SCD plays an important role in cellular cholesterol homeostasis and also in the secretion processes of triacylglycerol and phospholipid. Inhibition of SCD could be a useful treatment for metabollic disorders such as obesity and hepatic steatosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig, Hamster
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR