Tisk…

Anti-SMC4/CAPC Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3876-100EA 9730 CZK
BIOV3876-100
Anti-SMC4/CAPC Rabbit Polyclonal Antibody
Protilátky
The SMC (structural maintenance of chromosomes) family of proteins form heterodimeric complexes that modulate sister chromatid cohesion and chromosome condensation for mitosis. The two distinct classes of SMC protein complexes are comprised of SMC1 (also designated SB1.8) with SMC3 (also designated hCAP and Bamacan), and SMC2 (also designated hCAP-E for chromosome-associated polypeptide E) with hCAP-C. The SMC2/hCAP-C complex is required for mitotic chromosome condensation and functions independently of the SMC1/SMC3 complex during the cell cycle. Both SMC2 and hCAP-C are expressed throughout the cell cycle in a variety of cell lines, including HeLa cells, Daudi B cells, Jurkat T cells, SK2 neuronal cells, and HepG2 liver cells.

Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: SMC4/CAPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR