Tisk…

Anti-Notch 1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3881-100EA 7780 CZK
BIOV3881-100
Anti-Notch 1 Rabbit Polyclonal Antibody
Notch proteins are transmembrane receptor that regluate cell fate decisions. Four Notch homologs have been identified in mammals namely Notch 1, Notch 2, Notch 3 and Notch 4. The membrane ligand for Notch includes Jagged 1, Jagged 2 and Delta. Notch 1 is a 270 kDa transmembrane receptor that can be activated by Delta (Dl) ligand. Activation of Notch 1 will result in the proteolytic cleavage of Notch 1 into a 120 kDa length protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Notch 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR