Tisk…

Anti-Sphingosine Kinase 2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3884-100EA 7780 CZK
BIOV3884-100
Anti-Sphingosine Kinase 2 Rabbit Polyclonal Antibody
Sphingosine Kinase (SphK) is a conserved lipid kinase that catalyzes the phosphorylation of sphingolipid sphingosine to sphingosine-1-phosphate (SIP; SIP receptors coupled to cell surface G protein to regulate cell growth, survival, motility and inflammatory responses. There are two types of SphK, SphK1 and SphK2. SphK1 is found in the cytosol and migrates to the membrane when activated. SphK1 has been associated with cell growth, prevention of apoptosis and cellular transformation. SphK2 is found mainly in the nucleus and in contrast to SphK1, it enhances the rate of apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Sphingosine Kinase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR