Tisk…

Anti-TFAM Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3885-100EA 6910 CZK
BIOV3885-100
Anti-TFAM Rabbit Polyclonal Antibody
Protilátky
Transcription factor A mitochondria (TFAM) is an important activator of mitochondrial DNA transcription and replication. Studies in mice have shown that TFAM is required to regulate the mitochondrial genome copy number and is important for embryonic development. TFAM has been associated with mitochondrial disorder which includes ocular myopathy and muscular wasting in humans. Kearns-Sayre syndrome was shown in mouse model when expression of the gene was eliminated by targeted disruption in heart and muscle cells.

  • Recommended Dilutions: Western Blot: 0.5-4µg/ml

Type: Primary
Antigen: TFAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR