Tisk…

Anti-Sp3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3887-100EA 6910 CZK
BIOV3887-100
Anti-Sp3 Rabbit Polyclonal Antibody
Protilátky
Sp transcription factor family has a highly conserved DNA-binding domain composed of zinc fingers. The Sp family is comprised of Sp1, Sp2, Sp3 and Sp4 that regulates transcription by binding to consensus GC- and GT-box regulatory elements in the target genes. Sp1, Sp3 and Sp4 have a high binding capacity for GC box while Sp2 binds weakly to the GT box. Sp3 has been shown to have to a bifunctional transcription factor that can either stimulates or represses the transcription of various genes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Sp3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR