Tisk…

Anti-Boris Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3888-100EA 8120 CZK
BIOV3888-100
Anti-Boris Rabbit Polyclonal Antibody
Brother of the regulator of imprinted sites (BORIS) is a transcription factor also known as CCCTC- binding factor like protein (CTCF;Boris is usually expressed at a high level in the testis which correlates with male germ cell development and is also abnormally activated in a broad range of human cancers. Boris can be found in the nucles as well as in the cytoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Boris
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR