Tisk…

Anti-OX40 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3890-100EA 6910 CZK
BIOV3890-100
Anti-OX40 Rabbit Polyclonal Antibody
Protilátky
OX40 is also known as CD134 is a member of the tumor necrosis factor receptor superfamily (TNFR;OX40 plays an important role in CD4+ and CD8+T-cells and B-cells response. Binding of OX40 to its ligand, OX40L provides co-stimulatory signal to effector T-cells without CD28 activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: OX40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR