Tisk…

Anti-DKK3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3893-100EA 6910 CZK
BIOV3893-100
Anti-DKK3 Rabbit Polyclonal Antibody
Protilátky
Xenopus Dickkopf (Dkk) was initially discovered as a Wnt antagonist that plays an important role in head formation. By far, four members of Dkk have been identified in mammals. Each Dkk molecule contains two conserved cysteine-rich domains which are separated by a 50-55 amino acid linker region.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DKK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR