Tisk…

Anti-BLAME Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3895-100EA 6910 CZK
BIOV3895-100
Anti-BLAME Rabbit Polyclonal Antibody
Protilátky
BLAME (B Lymphocyte Activator Macrophage Expressed) also called SLAMF8. BLAME is a type I transmembrane protein that belongs to the CD2 family of Ig superfamily receptors. BLAME is mostly expressed on macrophages and dendritic cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BLAME
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR