Tisk…

Anti-DPP10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3898-200EA 6910 CZK
BIOV3898-200
Anti-DPP10 Rabbit Polyclonal Antibody
Protilátky
Dipeptidyl peptidase 10 or DPP10 is a single-pass type II membrane protein that belongs to the S9B family of DPP serine proteases. The conserved serine residue which normally found in the serine proteases family is not present in DPP10. Thus, DPP10 has no detectable proteases activity as the active serine site is replaced by a glycine residue. DPP10 cleaves the terminal dipeptides from cytokines and chemokines and also facilitates the voltage-gated potassium channels. It is expressed mostly in the brain and is a potential target for asthma treatment.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DPP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR