Tisk…

Anti-Hemopexin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3899-200EA 6910 CZK
BIOV3899-200
Anti-Hemopexin Rabbit Polyclonal Antibody
Protilátky
Hemopexin (HPX) is a plasma protein that has the highest binding affinity for heme. HPX prevents heme-mediated oxidative stress and heme-bound iron loss by transporting heme to the liver for breakdown.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Hemopexin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR