Tisk…

Anti-LAMP2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3900-100EA 7190 CZK
BIOV3900-100
Anti-LAMP2 Rabbit Polyclonal Antibody
Protilátky
Lysosomal-associated membrane protein 2 (LAMP2) also known as CD107B (Cluster of Differentiation 107B) is a membrane glycoprotein. LAMP2 plays an important role in the normal function of lysosome which includes protecting lysosomal membrane from autodigestion, maintaining lysosome acidity, and lysosome cellular adhesion.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR