Tisk…
Anti-Relaxin-2 Rabbit Polyclonal Antibody
Dodavatel: BioVision
BIOV3903-200EA 7780 CZK
BIOV3903-200
Anti-Relaxin-2 Rabbit Polyclonal Antibody
Relaxin 2 is a peptide hormone produced by the copora lutea of the ovary during pregnancy in many mammalian species. It is secreted into the blood just before parturition to help facilitate the birth process. Like insulin, relaxin is a heterodimer of two peptide chains A and B that are covalently linked by disulfide bonds.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml
Type: Primary
Antigen: Relaxin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human