Tisk…

Anti-Rabex-5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3905-100EA 8430 CZK
BIOV3905-100
Anti-Rabex-5 Rabbit Polyclonal Antibody
Rabex-5, also called RabGEF1 and RAP1, was identified as a guanine nucleotide exchange factor (GEF) for Rab5, a member of the Ras superfamily of small Rab GTPases (1;Rabex-5 generates the GTP-bound active form of Rab5 and forms a tight association with its effector protein Rabaptin-5. This complex localizes to endosomal membranes where it functions as a key regulator of vesicular trafficking during early endocytosis. Rabex-5 is also monoubiquitinated and has ubiquitin ligase activity that regulates its recruitment to early endosomes. The conformational change between Rab5 GTP/GDP states is essential for its biological function as a rate limiting regulator at multiple steps during endocytosis. Through its control of endosomal trafficking and endocytosis, Rabex-5 has been shown to negatively regulate NGF-mediated neurite outgrowth as well as FcεRI-dependent mast cell activation.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: Rabex-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR