Tisk…

Anti-CCL2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6730-100EA 7780 CZK
BIOV6730-100
Anti-CCL2 Rabbit Polyclonal Antibody
CCL2 is one of several cytokine genes clustered on the q-arm of chromosome 17. It is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. CCL2 displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.

Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:10–1:50, Immunofluorescence: ~1:10–1:50, FACS: ~1:10-1:50.

Type: Primary
Antigen: CCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR