Tisk…

Anti-ALDH5A1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6752-100EA 6910 CZK
BIOV6752-100
Anti-ALDH5A1 Rabbit Polyclonal Antibody
Protilátky
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH5A1 (aldehyde dehydrogenase 5 family, member A1), also known as SSDH or SSADH, is a 535 amino acid protein that localizes to the mitochondria and belongs to the aldehyde dehydrogenase family. Expressed in a variety of tissues, including liver, heart, lung, brain, kidney and placenta, ALDH5A1 is required for gamma-aminobutyric acid (GABA) recycling from the synaptic cleft. Mutations of ALDH5A1 lead to succinate semialdehyde dehydrogenase deficiency (SSADH deficiency) that is characterized by severe ataxia and by mildly retarded psychomotor development.

Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:10-1:50, FACS: ~1:10–1:50.

Type: Primary
Antigen: ALDH5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR