Tisk…

Anti-Beta-Actin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3917-100EA 8430 CZK
BIOV3917-100
Anti-Beta-Actin Rabbit Polyclonal Antibody
Actin is expressed in all eukaryotic cells and is the major component of the cytoskeleton. At least six types of actin are present in mammalian tissues and fall into three classes. Alpha actin expression is limited to various types of muscle and it regulates contactile potentials for the muscle cells, whereas beta and gamma actin, also known as cytoplasmic actin, are predominantly expressed in nonmuscle cells, controlling cell structure and motility.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Beta-Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR