Tisk…

Anti-OTX1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3918-100EA 9080 CZK
BIOV3918-100
Anti-OTX1 Rabbit Polyclonal Antibody
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper brain and sensory organ development and can cause epilepsy.

Recommended Dilutions: ELISA: 1:1,000; Western Blot: 1: 100~500).

Type: Primary
Antigen: OTX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR