Tisk…

Anti-Influenza A Virus Hemagglutinin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3924-100EA 6910 CZK
BIOV3924-100
Anti-Influenza A Virus Hemagglutinin Rabbit Polyclonal Antibody
Protilátky
Influenza A viruses are surface glycoprotein, single stranded, segmented RNA viruses that are found in birds. Influenza A subtypes are based on the virus glycoproteins hemagglutinin
(HA) and neuraminidase (NA;There are 16 different HA antigens and nine different N antigens for influenza A virus that have been isolated from wild birds.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Influenza A Virus Hemagglutinin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR