Tisk…

Anti-PAK6 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3927-100EA 6910 CZK
BIOV3927-100
Anti-PAK6 Rabbit Polyclonal Antibody
Protilátky
The PAK (p21-activated kinase) family of serine/threonine kinases plays an important role in multiple cellular processes, including cytoskeletal reorganization, MAPK signaling, apoptotic signaling, etc. Several mechanisms that induce PAK activation have been reported. Binding of Rac/cdc42 to the CRIB (or PBD) domain at the N-terminal region of PAK causes autophosphorylation and conformational change of PAK. More recently identified members PAK4, PAK5 and PAK6 have lower sequence similarity with PAK1-3 in the regulatory N-terminal region.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig, Horse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR