Tisk…

Anti-DNA Binding Protein-7 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3933-100EA 6910 CZK
BIOV3933-100
Anti-DNA Binding Protein-7 Rabbit Polyclonal Antibody
Protilátky
DNA binding protein 7 (DBP-7) is also known as Chd7. Mutations in the Chd7 gene is believed to be a major implication of CHARGE syndrome. CHARGE syndrome is associated with anomalies such as malformations of the heart, inner ear, retina and cranial nerve defects.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DNA Binding Protein-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR