Tisk…

Anti-PRMT-5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3935-100EA 6910 CZK
BIOV3935-100
Anti-PRMT-5 Rabbit Polyclonal Antibody
Protilátky
Protein arginine N-methyltransferase 5 (PRMT-5) is a member of the PRMT family that localizes to the cytoplasm in many types of tissues. At least three types of PRMT enzymes have been identified that shows essential regulatory functions by methylation of key proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PRMT-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR